missing translation for 'onlineSavingsMsg'
Learn More

2-Fluorotoluene 99.0+%, TCI America™

Catalog No. p-7129455
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F003925ML 25 mL
F0039500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F003925ML Supplier TCI America Supplier No. F003925ML
Only null left

Chemical Identifiers

CAS 95-52-3
Molecular Formula C7H7F
Molecular Weight (g/mol) 110.131
MDL Number MFCD00000322
InChI Key MMZYCBHLNZVROM-UHFFFAOYSA-N
Synonym 2-fluorotoluene, o-fluorotoluene, benzene, 1-fluoro-2-methyl, toluene, o-fluoro, 1-methyl-2-fluorobenzene, unii-l55xkb3k7h, 1-fluoro-2-methyl-benzene, ortho-fluorotoluene, fluorotoluene, l55xkb3k7h
PubChem CID 7241
IUPAC Name 1-fluoro-2-methylbenzene
SMILES CC1=CC=CC=C1F

Specifications

Color Yellow
Boiling Point 114°C
Quantity 25 mL
UN Number 2388
Formula Weight 110.13
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Fluorotoluene
EINECSNumber (3)-0083
RTECSNumber XT2579000
TSCA Yes