missing translation for 'onlineSavingsMsg'
Learn More

2-Fluorotoluene, 99+%

CAS: 95-52-3 | C7H7F | 110.131 g/mol

Supplier:  Thermo Scientific Chemicals A1691236

 View more versions of this product

Catalog No. AAA1691236


Only null left
Explore available promotions

Description

Description

It is employed in organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is employed in organic synthesis.

Solubility
Not miscible or difficult to mix in water.

Notes
Store away from oxidizing agents. Keep the container tightly sealed and store in cool, dry condition in well ventilated place.
Specifications

Specifications

2-Fluorotoluene
-87°C
113°C to 114°C
Aromatic
1.473
500 g
1853362
2-fluorotoluene, o-fluorotoluene, benzene, 1-fluoro-2-methyl, toluene, o-fluoro, 1-methyl-2-fluorobenzene, unii-l55xkb3k7h, 1-fluoro-2-methyl-benzene, ortho-fluorotoluene, fluorotoluene, l55xkb3k7h
MMZYCBHLNZVROM-UHFFFAOYSA-N
1-fluoro-2-methylbenzene
7241
≥99%
95-52-3
1.002
12°C (53°F)
C7H7F
MFCD00000322
UN2388
14,4180
Not miscible or difficult to mix in water.
CC1=CC=CC=C1F
110.131
110.13

Chemical Identifiers

95-52-3
110.131
MMZYCBHLNZVROM-UHFFFAOYSA-N
7241
CC1=CC=CC=C1F
C7H7F
MFCD00000322
2-fluorotoluene, o-fluorotoluene, benzene, 1-fluoro-2-methyl, toluene, o-fluoro, 1-methyl-2-fluorobenzene, unii-l55xkb3k7h, 1-fluoro-2-methyl-benzene, ortho-fluorotoluene, fluorotoluene, l55xkb3k7h
1-fluoro-2-methylbenzene
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H225-H335
Highly flammable liquid and vapour.
May cause respiratory irritation.

P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c

H225-H315-H319-H332-H335

DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLUOROTOLUENES

EINECSNumber : 202-428-5

RTECSNumber : XT2579000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only