missing translation for 'onlineSavingsMsg'
Learn More

2-Fluorothiophenol, 97%

CAS: 2557-78-0 | C6H4FS | 127.16 g/mol

Supplier:  Thermo Scientific Chemicals B2499506

 View more versions of this product

Catalog No. AAB2499506


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

2-Fluorothiophenol
1.203
56°C (133°F)
C6H4FS
MFCD00041419
UN3071
Air Sensitive
WJTZZPVVTSDNJJ-UHFFFAOYSA-M
2-fluorobenzenethiol
520216
97%
2557-78-0
164°C
Stench
1.555
5 g
2039768
2-fluorothiophenol, o-fluorothiophenol, o-fluorobenzenethiol, 2-fluorobenzene-1-thiol, fluorothiophenol, o-fluoro thiophenol, 2-fluoro-thiophenol, 2-fluoro-benzenethiol, 2-fluorobenzene thiol, pubchem5428
FC1=CC=CC=C1[S-]
127.16
128.17

Chemical Identifiers

2557-78-0
127.16
WJTZZPVVTSDNJJ-UHFFFAOYSA-M
520216
C6H4FS
MFCD00041419
2-fluorothiophenol, o-fluorothiophenol, o-fluorobenzenethiol, 2-fluorobenzene-1-thiol, fluorothiophenol, o-fluoro thiophenol, 2-fluoro-thiophenol, 2-fluoro-benzenethiol, 2-fluorobenzene thiol, pubchem5428
FC1=CC=CC=C1[S-]
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H300-H310-H331-H226-H315-H319-H335
Fatal if swallowed.
Fatal in contact with skin.
Toxic if inhaled.
Flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P210-P233-P235-P240-P241-P242-P243-P261-P264b-P270-P271-P280-P301+P310-P303+P361+P353-P304+P340-P305+P351+P338-P311-P312-P330-P332+P313-P363-P370+P378q-P501c

H226-H301+H311+H331-H315-H319-H335

DOTInformation : Hazard Class: 3; Packaging Group: III

EINECSNumber : 000-000-0

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only