missing translation for 'onlineSavingsMsg'
Learn More

2-Fluorothiophenol, 97%

Catalog No. AAB2499514
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2499514 25 g
AAB2499503 1 g
AAB2499506 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB2499514 Supplier Thermo Scientific Chemicals Supplier No. B2499514
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 2557-78-0 | C6H4FS | 127.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2557-78-0
Molecular Formula C6H4FS
Molecular Weight (g/mol) 127.16
MDL Number MFCD00041419
InChI Key WJTZZPVVTSDNJJ-UHFFFAOYSA-M
Synonym 2-fluorothiophenol, o-fluorothiophenol, o-fluorobenzenethiol, 2-fluorobenzene-1-thiol, fluorothiophenol, o-fluoro thiophenol, 2-fluoro-thiophenol, 2-fluoro-benzenethiol, 2-fluorobenzene thiol, pubchem5428
PubChem CID 520216
SMILES FC1=CC=CC=C1[S-]

Specifications

Density 1.203
Boiling Point 164°C
Flash Point 56°C (133°F)
Odor Stench
Refractive Index 1.555
Quantity 25 g
UN Number UN3071
Beilstein 2039768
Sensitivity Air Sensitive
IUPAC Name (2-fluorophenyl)sulfanide
Formula Weight 128.17
Percent Purity 97%
Chemical Name or Material 2-Fluorothiophenol
Show More Show Less
Hazard Category H226-H301+H311+H331-H315-H319-H335
Hazard Statement GHS H Statement
H300-H310-H331-H226-H315-H319-H335
Fatal if swallowed.
Fatal in contact with skin.
Toxic if inhaled.
Flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P270-P271-P280-P301+P310-P303+P361+P353-P304+P340-P305+P351+P338-P311-P312-P330-P332+P313-P363-P370+P378q-P501c
DOTInformation Hazard Class: 3; Packaging Group: III
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only