missing translation for 'onlineSavingsMsg'
Learn More

2-Fluorostyrene, 98%, stab. with 0.1% 4-tert-butylcatechol

Catalog No. AAA1742522
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1742522 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAA1742522 Supplier Thermo Scientific Chemicals Supplier No. A1742522
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 394-46-7 | C8H7F | 122.142 g/mol

It finds its application in the homo- and cross-olefin metathesis coupling of vinylphosphane oxides and electron-poor alkenes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It finds its application in the homo- and cross-olefin metathesis coupling of vinylphosphane oxides and electron-poor alkenes.

Solubility
Not miscible or difficult to mix in water.

Notes
Store away from oxidizing agents, heat, light, UV radiation and free radical initiators. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store in dark.

Chemical Identifiers

CAS 394-46-7
Molecular Formula C8H7F
Molecular Weight (g/mol) 122.142
MDL Number MFCD00013550
InChI Key YNQXOOPPJWSXMW-UHFFFAOYSA-N
Synonym 2-fluorostyrene, o-fluorostyrene, 1-fluoro-2-vinylbenzene, benzene, 1-ethenyl-2-fluoro, benzene, ethenylfluoro, 1-ethenyl-2-fluoro-benzene, 2-fluoro-styrene, acmc-1cuju, 1-fluoro-2-vinyl-benzene
PubChem CID 123056
IUPAC Name 1-ethenyl-2-fluorobenzene
SMILES C=CC1=CC=CC=C1F

Specifications

Density 1.02
Boiling Point 29°C to 30°C (4 mmHg)
Flash Point 35°C (95°F)
Odor Recognizable
Refractive Index 1.52
Quantity 100 g
UN Number UN1993
Beilstein 1925226
Solubility Information Not miscible or difficult to mix in water.
Formula Weight 122.14
Percent Purity 98%
Chemical Name or Material 2-Fluorostyrene, stabilized with 0.1% 4-tert-butylcatechol
Show More Show Less
Hazard Category H226-H315-H319-H335
Hazard Statement GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P233-P240-P243-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P374-P380-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
EINECSNumber 206-896-1
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only