missing translation for 'onlineSavingsMsg'
Learn More

2-Fluorophenylurea, 98%

Catalog No. AAL0124714
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0124714 25 g
AAL0124706 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0124714 Supplier Thermo Scientific Chemicals Supplier No. L0124714
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 656-31-5 | C7H7FN2O | 154.144 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 656-31-5
Molecular Formula C7H7FN2O
Molecular Weight (g/mol) 154.144
MDL Number MFCD00014786
InChI Key PAWVOCWEWJXILY-UHFFFAOYSA-N
Synonym 2-fluorophenyl urea, 1-2-fluorophenyl urea, o-fluorophenylurea, n-2-fluorophenyl urea, amino-n-2-fluorophenyl amide, o-fluorophenyl urea, acmc-20anu9, urea, o-fluorophenyl, urea,n-2-fluorophenyl
PubChem CID 12606
IUPAC Name (2-fluorophenyl)urea
SMILES C1=CC=C(C(=C1)NC(=O)N)F

Specifications

Melting Point 178°C to 184°C
Quantity 25 g
Beilstein 2803651
Formula Weight 154.14
Percent Purity 98%
Chemical Name or Material 2-Fluorophenylurea
EINECSNumber 211-513-6
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only