missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Fluorophenylacetic Acid 98.0+%, TCI America™
Supplier: TCI America F020525G
Specifications
| 2-Fluorophenylacetic Acid | |
| 63°C | |
| C8H7FO2 | |
| 25 g | |
| RPTRFSADOICSSK-UHFFFAOYSA-N | |
| 2-(2-fluorophenyl)acetic acid | |
| 67979 | |
| ≥98.0% (GC,T) |
| 451-82-1 | |
| White-Yellow | |
| MFCD00004315 | |
| 2-fluorophenylacetic acid, 2-2-fluorophenyl acetic acid, o-fluorophenylacetic acid, benzeneacetic acid, 2-fluoro, 2-fluorophenyl acetic acid, 2-fluorophenylaceticacid, acetic acid, o-fluorophenyl, pubchem4164, 2fluorophenylacetic acid | |
| C1=CC=C(C(=C1)CC(=O)O)F | |
| 154.14 | |
| 154.14 | |
| Crystalline Powder |
Chemical Identifiers
| 451-82-1 | |
| 154.14 | |
| RPTRFSADOICSSK-UHFFFAOYSA-N | |
| 67979 | |
| C1=CC=C(C(=C1)CC(=O)O)F |
| C8H7FO2 | |
| MFCD00004315 | |
| 2-fluorophenylacetic acid, 2-2-fluorophenyl acetic acid, o-fluorophenylacetic acid, benzeneacetic acid, 2-fluoro, 2-fluorophenyl acetic acid, 2-fluorophenylaceticacid, acetic acid, o-fluorophenyl, pubchem4164, 2fluorophenylacetic acid | |
| 2-(2-fluorophenyl)acetic acid |
Safety and Handling
TSCA : Yes