missing translation for 'onlineSavingsMsg'
Learn More

2-Fluorophenylacetic Acid 98.0+%, TCI America™

Catalog No. F02055G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F02055G 5 g
F020525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F02055G Supplier TCI America Supplier No. F02055G

Chemical Identifiers

CAS 451-82-1
Molecular Formula C8H7FO2
Molecular Weight (g/mol) 154.14
MDL Number MFCD00004315
InChI Key RPTRFSADOICSSK-UHFFFAOYSA-N
Synonym 2-fluorophenylacetic acid, 2-2-fluorophenyl acetic acid, o-fluorophenylacetic acid, benzeneacetic acid, 2-fluoro, 2-fluorophenyl acetic acid, 2-fluorophenylaceticacid, acetic acid, o-fluorophenyl, pubchem4164, 2fluorophenylacetic acid
PubChem CID 67979
IUPAC Name 2-(2-fluorophenyl)acetic acid
SMILES C1=CC=C(C(=C1)CC(=O)O)F

Specifications

Melting Point 63°C
Color White-Yellow
Quantity 5 g
Formula Weight 154.14
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Fluorophenylacetic Acid