missing translation for 'onlineSavingsMsg'
Learn More

2-Fluorophenylacetic acid, 98+%

Catalog No. p-7080866
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1059609 10 g
AAA1059618 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1059609 Supplier Thermo Scientific Chemicals Supplier No. A1059609
Only null left

CAS: 451-82-1 | C8H7FO2 | 154.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 451-82-1
Molecular Formula C8H7FO2
Molecular Weight (g/mol) 154.14
MDL Number MFCD00004315
InChI Key RPTRFSADOICSSK-UHFFFAOYSA-N
Synonym 2-fluorophenylacetic acid, 2-2-fluorophenyl acetic acid, o-fluorophenylacetic acid, benzeneacetic acid, 2-fluoro, 2-fluorophenyl acetic acid, 2-fluorophenylaceticacid, acetic acid, o-fluorophenyl, pubchem4164, 2fluorophenylacetic acid
PubChem CID 67979
IUPAC Name 2-(2-fluorophenyl)acetic acid
SMILES C1=CC=C(C(=C1)CC(=O)O)F

Specifications

Melting Point 61°C to 65°C
Odor Odorless
Quantity 10 g
Beilstein 775955
Formula Weight 154.14
Percent Purity ≥98%
Chemical Name or Material 2-Fluorophenylacetic acid
Hazard Category Warning
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
EINECSNumber 207-196-9
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only