missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Fluorophenyl Isocyanate 97.0+%, TCI America™
Supplier: TCI America I033825G
Specifications
| 2-Fluorophenyl Isocyanate | |
| Yellow | |
| C7H4FNO | |
| 25 g | |
| 2-fluorophenyl isocyanate, o-fluorophenyl isocyanate, 2-fluorophenylisocyanate, benzene, 1-fluoro-2-isocyanato, isocyanic acid 2-fluorophenyl ester, 2-fluorobenzenisocyanate, paragos 331045, labotest-bb ltbb005297, pubchem4455, 2-flurophenylisocyanate | |
| C1=CC=C(C(=C1)N=C=O)F | |
| 137.113 | |
| 137.11 | |
| Liquid |
| 16744-98-2 | |
| 65°C | |
| MFCD00001996 | |
| 2478 | |
| VZNCSZQPNIEEMN-UHFFFAOYSA-N | |
| 1-fluoro-2-isocyanatobenzene | |
| 85588 | |
| ≥97.0% (GC) |
Chemical Identifiers
| 16744-98-2 | |
| 137.113 | |
| VZNCSZQPNIEEMN-UHFFFAOYSA-N | |
| 85588 | |
| C1=CC=C(C(=C1)N=C=O)F |
| C7H4FNO | |
| MFCD00001996 | |
| 2-fluorophenyl isocyanate, o-fluorophenyl isocyanate, 2-fluorophenylisocyanate, benzene, 1-fluoro-2-isocyanato, isocyanic acid 2-fluorophenyl ester, 2-fluorobenzenisocyanate, paragos 331045, labotest-bb ltbb005297, pubchem4455, 2-flurophenylisocyanate | |
| 1-fluoro-2-isocyanatobenzene |
Safety and Handling
TSCA : No
Recommended Storage : Refrigerator