missing translation for 'onlineSavingsMsg'
Learn More

2-Fluorophenyl isocyanate, 98%

Catalog No. AAA1746606
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1746606 5 g
AAA1746614 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1746606 Supplier Thermo Scientific Chemicals Supplier No. A1746606
Only null left

CAS: 16744-98-2 | C7H4FNO | 137.113 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 16744-98-2
Molecular Formula C7H4FNO
Molecular Weight (g/mol) 137.113
MDL Number MFCD00001996
InChI Key VZNCSZQPNIEEMN-UHFFFAOYSA-N
Synonym 2-fluorophenyl isocyanate, o-fluorophenyl isocyanate, 2-fluorophenylisocyanate, benzene, 1-fluoro-2-isocyanato, isocyanic acid 2-fluorophenyl ester, 2-fluorobenzenisocyanate, paragos 331045, labotest-bb ltbb005297, pubchem4455, 2-flurophenylisocyanate
PubChem CID 85588
IUPAC Name 1-fluoro-2-isocyanatobenzene
SMILES C1=CC=C(C(=C1)N=C=O)F

Specifications

Density 1.221
Boiling Point 56°C to 58°C (10 mmHg)
Flash Point 50°C (122°F)
Odor Pungent, Makes Eyes Water
Refractive Index 1.5125
Quantity 5 g
UN Number UN3080
Beilstein 1210127
Sensitivity Moisture sensitive
Formula Weight 137.11
Percent Purity 98%
Chemical Name or Material 2-Fluorophenyl isocyanate
Show More Show Less
Hazard Category H226-H301+H311+H331-H315-H319-H334-H335
Hazard Statement GHS H Statement
H300-H330-H334-H226-H315-H319-H335
Fatal if swallowed.
Fatal if inhaled.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P270-P271-P280-P285-P301+P310-P303+P361+P353-P304+P340-P305+P351+P338-P311-P312-P330-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: ISOCYANATES, TOXIC, FLAMMABLE, N.O.S.
EINECSNumber 240-804-0
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only