missing translation for 'onlineSavingsMsg'
Learn More

2-Fluorophenethyl bromide, 97%

Catalog No. AC440120010
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC440120010 1 g
AC440120050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC440120010 Supplier Thermo Scientific Chemicals Supplier No. 440120010
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 91319-54-9 | C8H8BrF | 203.05 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 91319-54-9
Molecular Formula C8H8BrF
Molecular Weight (g/mol) 203.05
InChI Key FQGDFWIQBCQXPS-UHFFFAOYSA-N
Synonym 2-fluorophenethyl bromide, 1-2-bromoethyl-2-fluorobenzene, 1-2-bromo-ethyl-2-fluoro-benzene, benzene, 1-2-bromoethyl-2-fluoro, 2-2-bromoethyl-1-fluorobenzene, 1-2-bromoethyl-2-fluoro-benzene, 2-fluorophenyl-ethylbromide, acmc-209wk4, 2-fluorophenyl ethyl bromide, 2-2-fluorophenyl-ethylbromide
PubChem CID 2756964
IUPAC Name 1-(2-bromoethyl)-2-fluorobenzene
SMILES C1=CC=C(C(=C1)CCBr)F

Specifications

CAS Min % 96.0
CAS Max % 100.0
Color Colorless to Yellow
Boiling Point 73°C to 75°C (20.0 Torr)
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 203.05
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 2-Fluorophenethyl bromide
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Harmful if swallowed.
Causes serious eye irritation.
Toxic to aquatic life with long lasting effects.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Avoid release to the environment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only