missing translation for 'onlineSavingsMsg'
Learn More

2-Fluorocyclohexanol 98.0+%, TCI America™

Catalog No. p-7129796
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F06311G 1 g
F06315G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F06311G Supplier TCI America Supplier No. F06311G
Only null left

Chemical Identifiers

CAS 656-60-0
Molecular Formula C6H11FO
Molecular Weight (g/mol) 118.151
MDL Number MFCD08146648
InChI Key LMYKFDDTPIOYQV-PHDIDXHHSA-N
Synonym trans-2-fluorocyclohexanol, 1r,2r-2-fluorocyclohexan-1-ol, r,r---2-fluorocyclohexanol, trans-2-fluoro-cyclohexanol, 2alpha-fluorocyclohexan-1beta-ol, cyclohexanol, 2-fluoro-, 1r,2r, trans-2-fluorocyclohexanol unspecified stereochemistry
PubChem CID 12700773
IUPAC Name (1R,2R)-2-fluorocyclohexan-1-ol
SMILES C1CCC(C(C1)O)F

Specifications

Melting Point 21°C
Color White-Yellow
Boiling Point 166°C
Quantity 1 g
Formula Weight 118.15
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material 2-Fluorocyclohexanol
TSCA No
Recommended Storage Refrigerator