accessibility menu, dialog, popup

UserName

CAS RN 656-60-0

OHF

CAS RN 656-60-0

IUPAC Name: 2-fluorocyclohexan-1-ol
Synonyms: 2-fluorocyclohexan-1-ol
Molecular Weight (g/mol): 118.15
Molecular Formula: C6H11FO
InChi Key: LMYKFDDTPIOYQV-UHFFFAOYNA-N
SMILES: OC1CCCCC1F
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results