missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Fluorobiphenyl 97.0+%, TCI America™
Supplier: TCI America F026525G
Specifications
| 2-Fluorobiphenyl | |
| 74°C | |
| 248°C | |
| MFCD00000317 | |
| 2-fluorobiphenyl, 2-fluoro-1,1'-biphenyl, o-fluorodiphenyl, 1,1'-biphenyl, 2-fluoro, biphenyl, 2-fluoro, ortho-fluorodiphenyl, 2-fluorodiphenyl, 2-fluoro-biphenyl, ccris 1659, 2-fluoro-1-phenylbenzene | |
| FC1=CC=CC=C1C1=CC=CC=C1 | |
| 172.20 | |
| 172.20 | |
| Crystalline Powder |
| 321-60-8 | |
| White-Yellow | |
| C12H9F | |
| 25 g | |
| KLECYOQFQXJYBC-UHFFFAOYSA-N | |
| 2-fluoro-1,1'-biphenyl | |
| 67579 | |
| ≥97.0% (GC) |
Chemical Identifiers
| 321-60-8 | |
| 172.20 | |
| KLECYOQFQXJYBC-UHFFFAOYSA-N | |
| 67579 | |
| FC1=CC=CC=C1C1=CC=CC=C1 |
| C12H9F | |
| MFCD00000317 | |
| 2-fluorobiphenyl, 2-fluoro-1,1'-biphenyl, o-fluorodiphenyl, 1,1'-biphenyl, 2-fluoro, biphenyl, 2-fluoro, ortho-fluorodiphenyl, 2-fluorodiphenyl, 2-fluoro-biphenyl, ccris 1659, 2-fluoro-1-phenylbenzene | |
| 2-fluoro-1,1'-biphenyl |
Safety and Handling
TSCA : Yes