missing translation for 'onlineSavingsMsg'
Learn More

2-Fluorobiphenyl 97.0+%, TCI America™

Catalog No. p-7129581
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F02655G 5 g
F026525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F02655G Supplier TCI America Supplier No. F02655G
Only null left

Chemical Identifiers

CAS 321-60-8
Molecular Formula C12H9F
Molecular Weight (g/mol) 172.20
MDL Number MFCD00000317
InChI Key KLECYOQFQXJYBC-UHFFFAOYSA-N
Synonym 2-fluorobiphenyl, 2-fluoro-1,1'-biphenyl, o-fluorodiphenyl, 1,1'-biphenyl, 2-fluoro, biphenyl, 2-fluoro, ortho-fluorodiphenyl, 2-fluorodiphenyl, 2-fluoro-biphenyl, ccris 1659, 2-fluoro-1-phenylbenzene
PubChem CID 67579
IUPAC Name 2-fluoro-1,1'-biphenyl
SMILES FC1=CC=CC=C1C1=CC=CC=C1

Specifications

Melting Point 74°C
Color White-Yellow
Boiling Point 248°C
Quantity 5 g
Formula Weight 172.20
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Fluorobiphenyl