missing translation for 'onlineSavingsMsg'
Learn More

2-Fluorobenzylamine 98.0+%, TCI America™

Catalog No. p-7129739
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F05385G 5 g
F053825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F05385G Supplier TCI America Supplier No. F05385G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 89-99-6
Molecular Formula C7H8FN
Molecular Weight (g/mol) 125.146
InChI Key LRFWYBZWRQWZIM-UHFFFAOYSA-N
Synonym 2-fluorobenzylamine, 2-fluorophenyl methanamine, benzenemethanamine, 2-fluoro, o-fluorobenzylamine, benzylamine, o-fluoro, 1-2-fluorophenyl methanamine, 2-fluoro-benzylamine, 2-fluorobenzyl amine, 2-fluorophenyl methylamine, o-fluorobenzyl amine
PubChem CID 66649
IUPAC Name (2-fluorophenyl)methanamine
SMILES C1=CC=C(C(=C1)CN)F

Specifications

Color Yellow
Boiling Point 75°C
Quantity 5 g
UN Number 2735
Formula Weight 125.15
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 2-Fluorobenzylamine