missing translation for 'onlineSavingsMsg'
Learn More

2-Fluorobenzyl Chloride 98.0+%, TCI America™

Catalog No. F02115G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F02115G 5 g
F021125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F02115G Supplier TCI America Supplier No. F02115G

Chemical Identifiers

CAS 345-35-7
Molecular Formula C7H6ClF
Molecular Weight (g/mol) 144.57
MDL Number MFCD00000892
InChI Key MOBRMRJUKNQBMY-UHFFFAOYSA-N
Synonym 2-fluorobenzyl chloride, 1-chloromethyl-2-fluorobenzene, o-fluorobenzyl chloride, alpha-chloro-o-fluorotoluene, benzene, 1-chloromethyl-2-fluoro, alpha-chloro-2-fluorotoluene, 2-fluorobenzylchloride, 1-chloromethyl-2-fluoro-benzene, toluene, alpha-chloro-o-fluoro, o-fluorobenzylchloride
PubChem CID 9580
IUPAC Name 1-(chloromethyl)-2-fluorobenzene
SMILES FC1=CC=CC=C1CCl

Specifications

Color Yellow
Boiling Point 175°C
Quantity 5 g
UN Number 2920
Formula Weight 144.57
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Fluorobenzyl Chloride