missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Fluorobenzyl Alcohol 97.0+%, TCI America™
Supplier: TCI America F025925G
Specifications
| 2-Fluorobenzyl Alcohol | |
| Yellow | |
| C7H7FO | |
| 25 g | |
| QEHXDOJPVIHUDO-UHFFFAOYSA-N | |
| (2-fluorophenyl)methanol | |
| 67969 | |
| ≥97.0% (GC) |
| 446-51-5 | |
| 96°C | |
| MFCD00004601 | |
| 2-fluorobenzyl alcohol, 2-fluorophenyl methanol, o-fluorobenzyl alcohol, benzenemethanol, 2-fluoro, 2-fluorobenzylalcohol, 2-fluoro-benzenemethanol, 2-fluorobenzylic alcohol, 2-fluorophenyl methan-1-ol, pubchem3485, acmc-1asq8 | |
| OCC1=CC=CC=C1F | |
| 126.13 | |
| 126.13 | |
| Liquid |
Chemical Identifiers
| 446-51-5 | |
| 126.13 | |
| QEHXDOJPVIHUDO-UHFFFAOYSA-N | |
| 67969 | |
| OCC1=CC=CC=C1F |
| C7H7FO | |
| MFCD00004601 | |
| 2-fluorobenzyl alcohol, 2-fluorophenyl methanol, o-fluorobenzyl alcohol, benzenemethanol, 2-fluoro, 2-fluorobenzylalcohol, 2-fluoro-benzenemethanol, 2-fluorobenzylic alcohol, 2-fluorophenyl methan-1-ol, pubchem3485, acmc-1asq8 | |
| (2-fluorophenyl)methanol |
Safety and Handling
TSCA : No