missing translation for 'onlineSavingsMsg'
Learn More

2-Fluorobenzyl alcohol, 98%

Catalog No. AAA1479122
Change view
Click to view available options
Quantity:
10 g
50 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1479122 100 g
AAA1479109 10 g
AAA1479118 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1479122 Supplier Thermo Scientific Chemicals Supplier No. A1479122
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 446-51-5 | C7H7FO | 126.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 446-51-5
Molecular Formula C7H7FO
Molecular Weight (g/mol) 126.13
MDL Number MFCD00004601
InChI Key QEHXDOJPVIHUDO-UHFFFAOYSA-N
Synonym 2-fluorobenzyl alcohol, 2-fluorophenyl methanol, o-fluorobenzyl alcohol, benzenemethanol, 2-fluoro, 2-fluorobenzylalcohol, 2-fluoro-benzenemethanol, 2-fluorobenzylic alcohol, 2-fluorophenyl methan-1-ol, pubchem3485, acmc-1asq8
PubChem CID 67969
IUPAC Name (2-fluorophenyl)methanol
SMILES OCC1=CC=CC=C1F

Specifications

Density 1.175
Boiling Point 201°C
Flash Point 90°C (194°F)
Odor Characteristic
Refractive Index 1.514
Quantity 100 g
Beilstein 2079486
Formula Weight 126.13
Percent Purity 98%
Chemical Name or Material 2-Fluorobenzyl alcohol
Hazard Category H227-H315-H319
Precautionary Statement P210-P235-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
EINECSNumber 207-170-7
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only