missing translation for 'onlineSavingsMsg'
Learn More

2-Fluorobenzoyl Chloride 98.0+%, TCI America™

Catalog No. F019525G
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F019525G 25 g
F0195100G 100 g
F0195500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. F019525G Supplier TCI America Supplier No. F019525G

Chemical Identifiers

CAS 393-52-2
Molecular Formula C7H4ClFO
Molecular Weight (g/mol) 158.556
MDL Number MFCD00000656
InChI Key RAAGZOYMEQDCTD-UHFFFAOYSA-N
Synonym o-fluorobenzoyl chloride, benzoyl chloride, 2-fluoro, benzoyl chloride, o-fluoro, 2-fluorobenzoylchloride, 2-fluoro benzoyl chloride, unii-z4h017n76s, ccris 5914, fluorobenzoyl chloride, benzoyl chloride, fluoro, 2-fluorobenzoic acid chloride
PubChem CID 9808
IUPAC Name 2-fluorobenzoyl chloride
SMILES C1=CC=C(C(=C1)C(=O)Cl)F

Specifications

Color Yellow
Boiling Point 189°C
Quantity 25 g
UN Number 3265
Formula Weight 158.56
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 2-Fluorobenzoyl Chloride
RTECSNumber DM6640000
TSCA Yes