missing translation for 'onlineSavingsMsg'
Learn More

2-Fluorobenzoyl chloride, 97%

Catalog No. AAA1609622
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1609622 100 g
AAA1609636 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1609622 Supplier Thermo Scientific Chemicals Supplier No. A1609622

CAS: 393-52-2 | C7H4ClFO | 158.556 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 393-52-2
Molecular Formula C7H4ClFO
Molecular Weight (g/mol) 158.556
MDL Number MFCD00000656
InChI Key RAAGZOYMEQDCTD-UHFFFAOYSA-N
Synonym o-fluorobenzoyl chloride, benzoyl chloride, 2-fluoro, benzoyl chloride, o-fluoro, 2-fluorobenzoylchloride, 2-fluoro benzoyl chloride, unii-z4h017n76s, ccris 5914, fluorobenzoyl chloride, benzoyl chloride, fluoro, 2-fluorobenzoic acid chloride
PubChem CID 9808
IUPAC Name 2-fluorobenzoyl chloride
SMILES C1=CC=C(C(=C1)C(=O)Cl)F

Specifications

Melting Point 4°C to 6°C
Density 1.331
Boiling Point 207°C
Flash Point 82°C (179°F)
Odor Pungent
Refractive Index 1.5365
Quantity 100 g
UN Number UN3265
Beilstein 636864
Sensitivity Moisture Sensitive
Formula Weight 158.56
Percent Purity 97%
Chemical Name or Material 2-Fluorobenzoyl chloride
Show More Show Less
Hazard Category H227-H314
Hazard Statement GHS H Statement
H314-H318-H227
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.
Precautionary Statement P210-P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
EINECSNumber 206-887-2
RTECSNumber DM6640000
TSCA Yes

RUO – Research Use Only