Learn More
2-Fluorobenzotrifluoride, 99%
CAS: 392-85-8 | C7H4F4 | 164.103 g/mol
Supplier: Thermo Scientific Chemicals A1809506
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| 2-Fluorobenzotrifluoride | |
| -49°C | |
| 114°C to 115°C | |
| C7H4F4 | |
| MFCD00000374 | |
| UN1993 | |
| 2-fluorobenzotrifluoride, 1-fluoro-2-trifluoromethyl benzene, o-fluorobenzotrifluoride, benzene, 1-fluoro-2-trifluoromethyl, alpha,alpha,alpha,2-tetrafluorotoluene, benzene, fluoro trifluoromethyl, o,alpha,alpha,alpha-tetrafluorotoluene, a,a,a,2-tetrafluorotoluene, alpha,alpha,alpha,ar-tetrafluorotoluene, 2-trifluoromethyl fluorobenzene | |
| C1=CC=C(C(=C1)C(F)(F)F)F | |
| 164.103 | |
| 164.1 |
| 392-85-8 | |
| 1.293 | |
| 17°C (62°F) | |
| 1.406 | |
| 5 g | |
| 2046638 | |
| BGVGHYOIWIALFF-UHFFFAOYSA-N | |
| 1-fluoro-2-(trifluoromethyl)benzene | |
| 9807 | |
| 99% |
Chemical Identifiers
| 392-85-8 | |
| 164.103 | |
| BGVGHYOIWIALFF-UHFFFAOYSA-N | |
| 9807 | |
| C1=CC=C(C(=C1)C(F)(F)F)F |
| C7H4F4 | |
| MFCD00000374 | |
| 2-fluorobenzotrifluoride, 1-fluoro-2-trifluoromethyl benzene, o-fluorobenzotrifluoride, benzene, 1-fluoro-2-trifluoromethyl, alpha,alpha,alpha,2-tetrafluorotoluene, benzene, fluoro trifluoromethyl, o,alpha,alpha,alpha-tetrafluorotoluene, a,a,a,2-tetrafluorotoluene, alpha,alpha,alpha,ar-tetrafluorotoluene, 2-trifluoromethyl fluorobenzene | |
| 1-fluoro-2-(trifluoromethyl)benzene |
Safety and Handling
GHS H Statement
H225-H315-H319-H335
Highly flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
H225-H315-H319-H335
DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
EINECSNumber : 206-880-4
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only