missing translation for 'onlineSavingsMsg'
Learn More

2-Fluorobenzotrifluoride, 99%

Catalog No. AAA1809522
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1809522 100 g
AAA1809506 5 g
AAA1809514 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1809522 Supplier Thermo Scientific Chemicals Supplier No. A1809522
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 392-85-8 | C7H4F4 | 164.103 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 392-85-8
Molecular Formula C7H4F4
Molecular Weight (g/mol) 164.103
MDL Number MFCD00000374
InChI Key BGVGHYOIWIALFF-UHFFFAOYSA-N
Synonym 2-fluorobenzotrifluoride, 1-fluoro-2-trifluoromethyl benzene, o-fluorobenzotrifluoride, benzene, 1-fluoro-2-trifluoromethyl, alpha,alpha,alpha,2-tetrafluorotoluene, benzene, fluoro trifluoromethyl, o,alpha,alpha,alpha-tetrafluorotoluene, a,a,a,2-tetrafluorotoluene, alpha,alpha,alpha,ar-tetrafluorotoluene, 2-trifluoromethyl fluorobenzene
PubChem CID 9807
IUPAC Name 1-fluoro-2-(trifluoromethyl)benzene
SMILES C1=CC=C(C(=C1)C(F)(F)F)F

Specifications

Melting Point -49°C
Density 1.293
Boiling Point 114°C to 115°C
Flash Point 17°C (62°F)
Refractive Index 1.406
Quantity 100 g
UN Number UN1993
Beilstein 2046638
Formula Weight 164.1
Percent Purity 99%
Chemical Name or Material 2-Fluorobenzotrifluoride
Show More Show Less
Hazard Category H225-H315-H319-H335
Hazard Statement GHS H Statement
H225-H315-H319-H335
Highly flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
EINECSNumber 206-880-4
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only