missing translation for 'onlineSavingsMsg'
Learn More

2-Fluorobenzophenone 98.0+%, TCI America™

Catalog No. F03975G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F03975G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. F03975G Supplier TCI America Supplier No. F03975G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 342-24-5
Molecular Formula C13H9FO
Molecular Weight (g/mol) 200.212
MDL Number MFCD00000318
InChI Key DWFDQVMFSLLMPE-UHFFFAOYSA-N
Synonym 2-fluorobenzophenone, 2-fluorophenyl phenyl methanone, o-fluorobenzophenone, fluorobenzophenone, 2-fluorophenyl-phenylmethanone, 2-fluoro-phenyl-phenylmethanone, 2-fluorophenyl phenyl ketone, methanone, 2-fluorophenyl phenyl, pubchem4228, acmc-1ckt6
PubChem CID 67650
IUPAC Name (2-fluorophenyl)-phenylmethanone
SMILES C1=CC=C(C=C1)C(=O)C2=CC=CC=C2F

Specifications

Color Yellow
Boiling Point 190°C
Quantity 5 g
Formula Weight 200.21
Physical Form Liquid
Chemical Name or Material 2-Fluorobenzophenone