missing translation for 'onlineSavingsMsg'
Learn More

2-Fluorobenzonitrile, 99%

Catalog No. AAA1437509
Change view
Click to view available options
Quantity:
10 g
50 g
100 g
250 g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1437509 10 g
AAA1437518 50 g
AAA1437522 100 g
AAA1437530 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AAA1437509 Supplier Thermo Scientific Chemicals Supplier No. A1437509
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 394-47-8 | C7H4FN | 121.11 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 394-47-8
Molecular Formula C7H4FN
Molecular Weight (g/mol) 121.11
MDL Number MFCD00001773
InChI Key GDHXJNRAJRCGMX-UHFFFAOYSA-N
Synonym o-fluorobenzonitrile, benzonitrile, 2-fluoro, o-cyanofluorobenzene, 2-fluorobenzonitril, 2-cyanofluorobenzene, benzonitrile, o-fluoro, fluorobenzonitrile, 2-fluorobenzenecarbonitrile, 2-fluoro-benzonitrile, 1-cyano-2-fluorobenzene
PubChem CID 67855
IUPAC Name 2-fluorobenzonitrile
SMILES FC1=CC=CC=C1C#N

Specifications

Density 1.138
Boiling Point 196°C to 198°C
Flash Point 73°C (163°F)
Odor Almond-like
Refractive Index 1.507
Quantity 10 g
Beilstein 2042184
Formula Weight 121.11
Percent Purity 99%
Chemical Name or Material 2-Fluorobenzonitrile
Hazard Category H227-H302+H312+H332-H315-H318-H335
Hazard Statement GHS H Statement
H302-H312-H315-H319-H335-H227
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
Precautionary Statement P210-P235-P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P310-P312-P330-P332+P313-P362-P370+P378q-P501c
DOTInformation Hazard Class: 6.1; Packaging Group: III
EINECSNumber 206-897-7
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only