missing translation for 'onlineSavingsMsg'
Learn More

2-Fluorobenzohydrazide 98.0+%, TCI America™

Catalog No. F06165G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F06165G 5 g
F061625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F06165G Supplier TCI America Supplier No. F06165G

Chemical Identifiers

CAS 446-24-2
Molecular Formula C7H7FN2O
Molecular Weight (g/mol) 154.14
MDL Number MFCD00025112
InChI Key YJCCKQQVXNNAAR-UHFFFAOYSA-N
Synonym 2-fluorobenzhydrazide, 2-fluorobenzoic hydrazide, o-fluorobenzhydrazide, 2-fluorobenzoic acid hydrazide, 2-fluorobenzoylhydrazine, 2-fluorobenzenecarbohydrazide, pubchem3481, 2-fluoro-benzohydrazide, acmc-209jyw, 2-fluoranylbenzohydrazide
PubChem CID 136288
IUPAC Name 2-fluorobenzohydrazide
SMILES NNC(=O)C1=CC=CC=C1F

Specifications

Melting Point 73°C
Quantity 5 g
Formula Weight 154.14
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Fluorobenzohydrazide