missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Fluorobenzenethiol 98.0+%, TCI America™
$105.89 - $415.84
Chemical Identifiers
| CAS | 2557-78-0 |
|---|---|
| Molecular Formula | C6H4FS |
| Molecular Weight (g/mol) | 127.16 |
| MDL Number | MFCD00041419 |
| InChI Key | WJTZZPVVTSDNJJ-UHFFFAOYSA-M |
| Synonym | 2-fluorothiophenol, o-fluorothiophenol, o-fluorobenzenethiol, 2-fluorobenzene-1-thiol, fluorothiophenol, o-fluoro thiophenol, 2-fluoro-thiophenol, 2-fluoro-benzenethiol, 2-fluorobenzene thiol, pubchem5428 |
| PubChem CID | 520216 |
| IUPAC Name | (2-fluorophenyl)sulfanide |
| SMILES | FC1=CC=CC=C1[S-] |
Chemical Identifiers
| 2557-78-0 | |
| 127.16 | |
| WJTZZPVVTSDNJJ-UHFFFAOYSA-M | |
| 520216 | |
| FC1=CC=CC=C1[S-] |
| C6H4FS | |
| MFCD00041419 | |
| 2-fluorothiophenol, o-fluorothiophenol, o-fluorobenzenethiol, 2-fluorobenzene-1-thiol, fluorothiophenol, o-fluoro thiophenol, 2-fluoro-thiophenol, 2-fluoro-benzenethiol, 2-fluorobenzene thiol, pubchem5428 | |
| (2-fluorophenyl)sulfanide |
Specifications
| 2557-78-0 | |
| 164°C | |
| MFCD00041419 | |
| 3336 | |
| WJTZZPVVTSDNJJ-UHFFFAOYSA-M | |
| (2-fluorophenyl)sulfanide | |
| 520216 | |
| ≥98.0% (GC) | |
| 2-Fluorobenzenethiol |
| Yellow | |
| C6H4FS | |
| 5 g | |
| 2-fluorothiophenol, o-fluorothiophenol, o-fluorobenzenethiol, 2-fluorobenzene-1-thiol, fluorothiophenol, o-fluoro thiophenol, 2-fluoro-thiophenol, 2-fluoro-benzenethiol, 2-fluorobenzene thiol, pubchem5428 | |
| FC1=CC=CC=C1[S-] | |
| 127.16 | |
| 128.16 | |
| Liquid |
Safety and Handling
TSCA : No