missing translation for 'onlineSavingsMsg'
Learn More

2-Fluoroanisole 96.0+%, TCI America™

Catalog No. p-7129556
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F023025ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. F023025ML Supplier TCI America Supplier No. F023025ML

Chemical Identifiers

CAS 321-28-8
Molecular Formula C7H7FO
Molecular Weight (g/mol) 126.13
MDL Number MFCD00000316
InChI Key JIXDOBAQOWOUPA-UHFFFAOYSA-N
Synonym 2-fluoroanisole, o-fluoroanisole, benzene, 1-fluoro-2-methoxy, ortho-fluoroanisole, anisole, o-fluoro, 1-fluoro-2-methoxy-benzene, paragos 390201, timtec-bb sbb040818, fr bo1 wln, fluoroanisole
PubChem CID 67576
IUPAC Name 1-fluoro-2-methoxybenzene
SMILES COC1=CC=CC=C1F

Specifications

Melting Point -39°C
Color Yellow
Boiling Point 160°C
Quantity 25 mL
UN Number 1993
Formula Weight 126.13
Percent Purity ≥96.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Fluoroanisole