missing translation for 'onlineSavingsMsg'
Learn More

2-Fluoroaniline 99.0+%, TCI America™

Supplier:  TCI America F0032100G

 View more versions of this product

Catalog No. F0032100G


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

2-Fluoroaniline
170°C
MFCD00007642
2941
FTZQXOJYPFINKJ-UHFFFAOYSA-N
2-fluoroaniline
9584
111.12
Liquid
348-54-9
C6H6FN
100 g
o-fluoroaniline, benzenamine, 2-fluoro, 1-amino-2-fluorobenzene, 2-fluorobenzenamine, 2-fluoro-phenylamine, 2-fluoro-benzenamine, fluoroaniline-2, o-aminofluorobenzene, aniline, o-fluoro, 2-fluorophenylamine
NC1=CC=CC=C1F
111.12
CHEBI:27526
≥99.0% (GC)

Chemical Identifiers

348-54-9
111.12
FTZQXOJYPFINKJ-UHFFFAOYSA-N
9584
2-fluoroaniline
C6H6FN
MFCD00007642
o-fluoroaniline, benzenamine, 2-fluoro, 1-amino-2-fluorobenzene, 2-fluorobenzenamine, 2-fluoro-phenylamine, 2-fluoro-benzenamine, fluoroaniline-2, o-aminofluorobenzene, aniline, o-fluoro, 2-fluorophenylamine
CHEBI:27526
NC1=CC=CC=C1F
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : (3)-0222

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions