missing translation for 'onlineSavingsMsg'
Learn More

2-Fluoroaniline 99.0+%, TCI America™

Catalog No. F003225G
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F003225G 25 g
F0032100G 100 g
F0032500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. F003225G Supplier TCI America Supplier No. F003225G

Chemical Identifiers

CAS 348-54-9
Molecular Formula C6H6FN
Molecular Weight (g/mol) 111.12
MDL Number MFCD00007642
InChI Key FTZQXOJYPFINKJ-UHFFFAOYSA-N
Synonym o-fluoroaniline, benzenamine, 2-fluoro, 1-amino-2-fluorobenzene, 2-fluorobenzenamine, 2-fluoro-phenylamine, 2-fluoro-benzenamine, fluoroaniline-2, o-aminofluorobenzene, aniline, o-fluoro, 2-fluorophenylamine
PubChem CID 9584
ChEBI CHEBI:27526
IUPAC Name 2-fluoroaniline
SMILES NC1=CC=CC=C1F

Specifications

Boiling Point 170°C
Quantity 25 g
UN Number 2941
Formula Weight 111.12
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Fluoroaniline
EINECSNumber (3)-0222
TSCA Yes