missing translation for 'onlineSavingsMsg'
Learn More

2-Fluoroaniline, 99%

Catalog No. AAA1261236
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1261236 500 g
AAA1261214 25 g
AAA1261222 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1261236 Supplier Thermo Scientific Chemicals Supplier No. A1261236

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 348-54-9 | C6H6FN | 111.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 348-54-9
Molecular Formula C6H6FN
Molecular Weight (g/mol) 111.12
MDL Number MFCD00007642
InChI Key FTZQXOJYPFINKJ-UHFFFAOYSA-N
Synonym o-fluoroaniline, benzenamine, 2-fluoro, 1-amino-2-fluorobenzene, 2-fluorobenzenamine, 2-fluoro-phenylamine, 2-fluoro-benzenamine, fluoroaniline-2, o-aminofluorobenzene, aniline, o-fluoro, 2-fluorophenylamine
PubChem CID 9584
ChEBI CHEBI:27526
IUPAC Name 2-fluoroaniline
SMILES NC1=CC=CC=C1F

Specifications

Melting Point -29°C
Density 1.152
Boiling Point 171°C to 173°C
Flash Point 60°C (140°F)
Odor Recognizable
Refractive Index 1.542
Quantity 500 g
UN Number UN2941
Beilstein 1524219
Formula Weight 111.12
Percent Purity 99%
Chemical Name or Material 2-Fluoroaniline
Show More Show Less
Hazard Category H226-H302+H312+H332-H315-H318-H335
Hazard Statement GHS H Statement
H331-H226-H373-H302-H312-H315-H319
Toxic if inhaled.
Flammable liquid and vapour.
May cause damage to organs through prolonged or repeated exposure.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P270-P271-P280-P301+P312-P303+P361+P353-P304+P340-P305+P351+P338-P310-P312-P330-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: FLUOROANILINES
EINECSNumber 206-478-9
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only