missing translation for 'onlineSavingsMsg'
Learn More

2'-Fluoroacetophenone 97.0+%, TCI America™

Catalog No. p-7129638
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F03645G 5 g
F036425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F03645G Supplier TCI America Supplier No. F03645G
Only null left

Chemical Identifiers

CAS 445-27-2
Molecular Formula C8H7FO
Molecular Weight (g/mol) 138.141
MDL Number MFCD00000320
InChI Key QMATYTFXDIWACW-UHFFFAOYSA-N
Synonym 2'-fluoroacetophenone, 1-2-fluorophenyl ethanone, o-fluoroacetophenone, 1-2-fluorophenyl ethan-1-one, 1-acetyl-2-fluorobenzene, ethanone, 1-fluorophenyl, 2-fluoroacetophenone, 2'fluoroacetophenone, 2;-fluoroacetophenone
PubChem CID 96744
IUPAC Name 1-(2-fluorophenyl)ethanone
SMILES CC(=O)C1=CC=CC=C1F

Specifications

Color Yellow
Boiling Point 85°C
Quantity 5 g
Formula Weight 138.14
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 2′-Fluoroacetophenone