accessibility menu, dialog, popup

UserName

H3COF

CAS RN 445-27-2

IUPAC Name: 1-(2-fluorophenyl)ethan-1-one
Synonyms: 1-(2-fluorophenyl)ethanone
Molecular Weight (g/mol): 138.14
Molecular Formula: C8H7FO
InChi Key: QMATYTFXDIWACW-UHFFFAOYSA-N
SMILES: CC(=O)C1=CC=CC=C1F
Boiling Point 
  • (3)
  • (1)
  • (2)
Molecular Weight (g/mol) 
  • (1)
  • (5)
Percent Purity 
  • (2)
  • (4)
Quantity 
  • (1)
  • (3)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (2)
  • (4)
  • (3)
  • (1)
  • (2)
  • (1)
  • (5)
  • (2)
  • (4)
  • (1)
  • (3)
  • (2)
  • (3)