missing translation for 'onlineSavingsMsg'
Learn More

2'-Fluoroacetophenone, 97%

Catalog No. AAA1512806
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1512806 5 g
AAA1512814 25 g
AAA1512822 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1512806 Supplier Thermo Scientific Chemicals Supplier No. A1512806
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 445-27-2 | C8H7FO | 138.141 g/mol

2'-Fluoroacetophenone is used as starting reagent in the synthesis of ascididemin. It is used to produce 1-(2-piperidin-1-yl-phenyl)-ethanone by reaction with piperidine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2′-Fluoroacetophenone is used as starting reagent in the synthesis of ascididemin. It is used to produce 1-(2-piperidin-1-yl-phenyl)-ethanone by reaction with piperidine.

Solubility
Soluble in Acetone, Chloroform, Dichloromethane, Ethanol, Ethyl Acetate and Methanol.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.

Chemical Identifiers

CAS 445-27-2
Molecular Formula C8H7FO
Molecular Weight (g/mol) 138.141
MDL Number MFCD00000320
InChI Key QMATYTFXDIWACW-UHFFFAOYSA-N
Synonym 2'-fluoroacetophenone, 1-2-fluorophenyl ethanone, o-fluoroacetophenone, 1-2-fluorophenyl ethan-1-one, 1-acetyl-2-fluorobenzene, ethanone, 1-fluorophenyl, 2-fluoroacetophenone, 2'fluoroacetophenone, 2;-fluoroacetophenone
PubChem CID 96744
IUPAC Name 1-(2-fluorophenyl)ethanone
SMILES CC(=O)C1=CC=CC=C1F

Specifications

Density 1.137
Boiling Point 82°C to 83°C (10 mmHg)
Flash Point 61°C (141°F)
Refractive Index 1.5075
Quantity 5 g
Beilstein 2041344
Solubility Information Soluble in Acetone,Chloroform,Dichloromethane,Ethanol,Ethyl Acetate and Methanol.
Formula Weight 138.14
Percent Purity 97%
Chemical Name or Material 2'-Fluoroacetophenone
Hazard Category H227-H302-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
Precautionary Statement P210-P235-P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P370+P378q-P501c
EINECSNumber 207-156-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only