missing translation for 'onlineSavingsMsg'
Learn More

2-Fluoro-L-phenylalanine 96.0+%, TCI America™

Catalog No. F02731G
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F02731G 1 g
F0273100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F02731G Supplier TCI America Supplier No. F02731G
Only null left

Chemical Identifiers

CAS 19883-78-4
Molecular Formula C9H10FNO2
Molecular Weight (g/mol) 183.18
MDL Number MFCD00066448
InChI Key NYCRCTMDYITATC-QMMMGPOBSA-N
Synonym 2-fluoro-l-phenylalanine, h-phe 2-f-oh, l-2-fluorophenylalanine, 2s-2-amino-3-2-fluorophenyl propanoic acid, l-2-f-phe-oh, o-fluoro-l-phenylalanine, 2-fluoro-l-phe, phenylalanine, 2-fluoro, s-2-amino-3-2-fluorophenyl propanoic acid, l-2-fluorophe
PubChem CID 716319
IUPAC Name (2S)-2-amino-3-(2-fluorophenyl)propanoic acid
SMILES N[C@@H](CC1=CC=CC=C1F)C(O)=O

Specifications

Melting Point 217°C
Color White-Yellow
Quantity 1 g
Formula Weight 183.18
Percent Purity ≥96.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Fluoro-L-phenylalanine