accessibility menu, dialog, popup

UserName

CAS RN 19883-78-4

NH2(S)FOHO

CAS RN 19883-78-4

IUPAC Name: (2S)-2-amino-3-(2-fluorophenyl)propanoic acid
Synonyms: (2S)-2-amino-3-(2-fluorophenyl)propanoic acid
Molecular Weight (g/mol): 183.18
Molecular Formula: C9H10FNO2
InChi Key: NYCRCTMDYITATC-QMMMGPOBSA-N
SMILES: N[C@@H](CC1=CC=CC=C1F)C(O)=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results