missing translation for 'onlineSavingsMsg'
Learn More

2-Fluoro-beta-alanine Hydrochloride 99.0+%, TCI America™

Catalog No. F0185100MG
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F0185100MG 100 mg
F01851G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F0185100MG Supplier TCI America Supplier No. F0185100MG

Chemical Identifiers

CAS 867-84-5
Molecular Formula C3H7ClFNO2
Molecular Weight (g/mol) 143.54
MDL Number MFCD00060202
InChI Key CVYOZQFAVJXJTK-UHFFFAOYNA-N
Synonym 3-Amino-2-fluoropropionic Acid Hydrochloride
PubChem CID 44630296
IUPAC Name 2-carboxy-2-fluoroethan-1-aminium chloride
SMILES [Cl-].[NH3+]CC(F)C(O)=O

Specifications

Color White-Yellow
Quantity 100 mg
Formula Weight 143.54
Percent Purity ≥99.0% (N)
Grade Laboratory
Physical Form Crystalline Powder
Chemical Name or Material 2-Fluoro-beta-alanine Hydrochloride
RTECSNumber AY6200000
TSCA No