accessibility menu, dialog, popup

UserName

ClH3NFOHO

CAS RN 867-84-5

IUPAC Name: 2-carboxy-2-fluoroethan-1-aminium chloride
Synonyms: 2-carboxy-2-fluoroethanaminium chloride
Molecular Weight (g/mol): 143.54
Molecular Formula: C3H7ClFNO2
InChi Key: CVYOZQFAVJXJTK-UHFFFAOYNA-N
SMILES: [Cl-].[NH3+]CC(F)C(O)=O
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)
  • (1)