missing translation for 'onlineSavingsMsg'
Learn More

2-Fluoro-alpha-methyl-4-biphenylacetic acid, 99%

CAS: 5104-49-4 | C15H13FO2 | 244.27 g/mol

Supplier:  Thermo Scientific Chemicals B2260306

 View more versions of this product

Catalog No. AAB2260306


Only null left

Description

Description

A cyclooxygenase inhibitor

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

2-Fluoro-alpha-methyl-4-biphenylacetic acid
112°C to 114°C
C15H13FO2
5 g
14,4199
SYTBZMRGLBWNTM-UHFFFAOYNA-N
2-{2-fluoro-[1,1'-biphenyl]-4-yl}propanoic acid
3394
244.27
5104-49-4
Odorless
MFCD00079303
UN2811
flurbiprofen, ansaid, froben, antadys, cebutid, flurofen, anside, flurbiprofene, flurbiprofeno, flurbiprofenum
CC(C(O)=O)C1=CC=C(C(F)=C1)C1=CC=CC=C1
244.27
CHEBI:5130
99%

Chemical Identifiers

5104-49-4
244.27
SYTBZMRGLBWNTM-UHFFFAOYNA-N
3394
CC(C(O)=O)C1=CC=C(C(F)=C1)C1=CC=CC=C1
C15H13FO2
MFCD00079303
flurbiprofen, ansaid, froben, antadys, cebutid, flurofen, anside, flurbiprofene, flurbiprofeno, flurbiprofenum
CHEBI:5130
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H301-H315-H319-H335

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.

EINECSNumber : 225-827-6

RTECSNumber : DU8341000

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only