missing translation for 'onlineSavingsMsg'
Learn More

2-Fluoro-6-(trifluoromethyl)benzyl Bromide 98.0+%, TCI America™

Catalog No. F07325G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F07325G 5 g
F07321G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F07325G Supplier TCI America Supplier No. F07325G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 239087-08-2
Molecular Formula C8H5BrF4
Molecular Weight (g/mol) 257.026
MDL Number MFCD00082477
InChI Key RINUERVPFANASB-UHFFFAOYSA-N
Synonym 2-fluoro-6-trifluoromethyl benzyl bromide, 2-bromomethyl-1-fluoro-3-trifluoromethyl benzene, 2-fluoro-6-trifluoromethylbenzyl bromide, 2-fluoro-6-trifluoromethylbenzylbromide, 2-fluoro-6-trifluoromethyl benzylbromide, 2-bromomethyl-3-fluorobenzotrifluoride, alpha-bromo-2-fluoro-6-trifluoromethyl toluene, benzene, 2-bromomethyl-1-fluoro-3-trifluoromethyl, pubchem4932, acmc-1cg83
PubChem CID 2737573
IUPAC Name 2-(bromomethyl)-1-fluoro-3-(trifluoromethyl)benzene
SMILES C1=CC(=C(C(=C1)F)CBr)C(F)(F)F

Specifications

Melting Point 44°C
Color White-Yellow
Boiling Point 42°C
Quantity 5 g
UN Number 3261
Formula Weight 257.03
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Fluoro-6-(trifluoromethyl)benzyl Bromide