missing translation for 'onlineSavingsMsg'
Learn More

2-Fluoro-6-(trifluoromethyl)benzonitrile 98.0+%, TCI America™

Catalog No. F07015G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F07015G 5 g
F07011G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F07015G Supplier TCI America Supplier No. F07015G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 133116-83-3
Molecular Formula C8H3F4N
Molecular Weight (g/mol) 189.113
MDL Number MFCD00061153
InChI Key OGQYJDHTHFAPRN-UHFFFAOYSA-N
Synonym 2-fluoro-6-trifluoromethyl benzonitrile, 2-cyano-3-fluorobenzotrifluoride, 2-fluoro-6-trifluoromethyl-benzonitrile, benzonitrile, 2-fluoro-6-trifluoromethyl, .alpha.,.alpha.,.alpha.,6-tetrafluoro-o-tolunitrile, pubchem4799, acmc-1cgr6, intermediates-zcf02031, ksc495m8r
PubChem CID 145613
IUPAC Name 2-fluoro-6-(trifluoromethyl)benzonitrile
SMILES C1=CC(=C(C(=C1)F)C#N)C(F)(F)F

Specifications

Melting Point 28°C
Color White-Yellow
Quantity 5 g
UN Number 3439
Formula Weight 189.11
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material 2-Fluoro-6-(trifluoromethyl)benzonitrile