missing translation for 'onlineSavingsMsg'
Learn More

2-Fluoro-6-nitrotoluene 98.0+%, TCI America™

Catalog No. p-7129837
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F06965G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. F06965G Supplier TCI America Supplier No. F06965G

Chemical Identifiers

CAS 769-10-8
Molecular Formula C7H6FNO2
Molecular Weight (g/mol) 155.13
MDL Number MFCD00007197
InChI Key GXPIVRKDWZKIKZ-UHFFFAOYSA-N
Synonym 2-fluoro-6-nitrotoluene, 1-fluor-2-methyl-3-nitrobenzene, 2-nitro-6-fluorotoluene, 6-fluoro-2-nitrotoluene, benzene, 1-fluoro-2-methyl-3-nitro, 3-fluoro-2-methyl-1-nitrobenzene, 1-fluor-2-methyl-3-nitrobenzol, pubchem1594, acmc-1bc1k
PubChem CID 69854
IUPAC Name 1-fluoro-2-methyl-3-nitrobenzene
SMILES CC1=C(F)C=CC=C1[N+]([O-])=O

Specifications

Melting Point 7°C
Color Yellow
Boiling Point 97°C
Quantity 5 g
Formula Weight 155.13
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Fluoro-6-nitrotoluene