missing translation for 'onlineSavingsMsg'
Learn More

2-Fluoro-6-nitrophenol 98.0+%, TCI America™

Catalog No. F06231G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F06231G 1 g
F06235G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F06231G Supplier TCI America Supplier No. F06231G

Chemical Identifiers

CAS 1526-17-6
Molecular Formula C6H4FNO3
Molecular Weight (g/mol) 157.10
MDL Number MFCD00042446
InChI Key HIGRXCJEFUYRNW-UHFFFAOYSA-N
Synonym 2-fluoro-6-nitrophenol, 6-fluoro-2-nitrophenol, phenol, 2-fluoro-6-nitro, 2-nitro-6-fluorophenol, 2-fluoro-6-nitro-phenol, 3-fluoro-2-hydroxynitrobenzene, fluoro-6-nitrophenol, 2, 2-fluoro-6-nitrophen, pubchem10654, 2-fluor-6-nitrophenol
PubChem CID 73710
IUPAC Name 2-fluoro-6-nitrophenol
SMILES OC1=C(F)C=CC=C1[N+]([O-])=O

Specifications

Melting Point 92°C
Color Yellow
Quantity 1 g
Formula Weight 157.10
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Fluoro-6-nitrophenol