missing translation for 'onlineSavingsMsg'
Learn More

2-Fluoro-6-nitrobenzoic acid, 98%

Catalog No. p-7054640
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6301306 5 g
AAH6301314 25 g
AAH6301322 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH6301306 Supplier Thermo Scientific Chemicals Supplier No. H6301306

CAS: 385-02-4 | C7H4FNO4 | 185.11 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 385-02-4
Molecular Formula C7H4FNO4
Molecular Weight (g/mol) 185.11
MDL Number MFCD01862079
InChI Key MPDZCNPDHUUPRL-UHFFFAOYSA-N
Synonym 2-fluoro-6-nitrobenzoic acid, 2-fluoro-6-nitrobenzoicacid, 6-fluoro-2-nitrobenzoic acid, 2-carboxy-3-fluoronitrobenzene, 2-fluoro-6-nitro-benzoic acid, benzoic acid, 2-fluoro-6-nitro, pubchem1308, 6-fluoro-nitrobenzoic acid, acmc-209j0j, ksc222c6b
PubChem CID 603758
SMILES C1=CC(=C(C(=C1)F)C(=O)O)[N+](=O)[O-]

Specifications

Quantity 5 g
Formula Weight 185.11
Percent Purity 98%
Physical Form Solid
Chemical Name or Material 2-Fluoro-6-nitrobenzoic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only