missing translation for 'onlineSavingsMsg'
Learn More

2-Fluoro-6-methoxybenzamide, 98%

Catalog No. AAH32774MD
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH32774MD 250 mg
AAH3277403 1 g
AAH3277406 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH32774MD Supplier Thermo Scientific Chemicals Supplier No. H32774MD

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 529512-81-0 | C8H8FNO2 | 169.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 529512-81-0
Molecular Formula C8H8FNO2
Molecular Weight (g/mol) 169.16
MDL Number MFCD00277474
InChI Key BDJNFURQDZEPLK-UHFFFAOYSA-N
Synonym 2-fluoro-6-methoxy-benzamide, 2-methoxy-6-fluorobenzamide, 2-fluoro-6-methoxy benzamide, #, benzamide, 2-fluoro-6-methoxy
PubChem CID 579738
IUPAC Name 2-fluoro-6-methoxybenzamide
SMILES COC1=C(C(N)=O)C(F)=CC=C1

Specifications

Melting Point 190°C to 194°C
Quantity 250 mg
Formula Weight 169.15
Percent Purity 98%
Chemical Name or Material 2-Fluoro-6-methoxybenzamide
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only