missing translation for 'onlineSavingsMsg'
Learn More

2-Fluoro-6-iodobenzonitrile 98.0+%, TCI America™

Catalog No. p-7129817
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F06601G 1 g
F06605G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F06601G Supplier TCI America Supplier No. F06601G
Only null left

Chemical Identifiers

CAS 79544-29-9
Molecular Formula C7H3FIN
Molecular Weight (g/mol) 247.01
MDL Number MFCD00015478
InChI Key FAACTMVXBNSPJA-UHFFFAOYSA-N
Synonym 2-iodo-6-fluorobenzonitrile, 2-cyano-3-fluoro-1-iodobenzene, benzonitrile, 2-fluoro-6-iodo, 6-fluoro-2-iodobenzenecarbonitrile, 2-fluoro-6-iodo-benzonitrile, 6-iodo-2-fluorobenzonitrile, pubchem4780, acmc-209php, ksc494e4h, pharmabridge p-2313
PubChem CID 522722
IUPAC Name 2-fluoro-6-iodobenzonitrile
SMILES FC1=CC=CC(I)=C1C#N

Specifications

Melting Point 53°C
Color White-Yellow
Quantity 1 g
UN Number 3439
Formula Weight 247.01
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Fluoro-6-iodobenzonitrile