missing translation for 'onlineSavingsMsg'
Learn More

2-Fluoro-6-iodobenzoic Acid 98.0+%, TCI America™

Catalog No. F07961G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F07961G 1 g
F07965G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F07961G Supplier TCI America Supplier No. F07961G

Chemical Identifiers

CAS 111771-08-5
Molecular Formula C7H4FIO2
Molecular Weight (g/mol) 266.01
MDL Number MFCD00042289
InChI Key CYCXAPWOBWWNRK-UHFFFAOYSA-N
Synonym 6-fluoro-2-iodobenzoic acid, benzoic acid, 2-fluoro-6-iodo, maybridge3_007452, pubchem1386, acmc-2099an, 2-fluoro-6-iodobenzoicacid, 2-fluoro-6-iodo benzoic acid, 2-fluoro-6-iodo-benzoic acid, rarechem al bo 0280, attercop-chm at126295
PubChem CID 2733302
IUPAC Name 2-fluoro-6-iodobenzoic acid
SMILES C1=CC(=C(C(=C1)I)C(=O)O)F

Specifications

Melting Point 126°C
Quantity 1 g
Formula Weight 266.01
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Fluoro-6-iodobenzoic Acid