missing translation for 'onlineSavingsMsg'
Learn More

2-Fluoro-6-iodobenzoic acid, 97%

Catalog No. p-7044715
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0792403 1 g
AAL0792406 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0792403 Supplier Thermo Scientific Chemicals Supplier No. L0792403
Only null left

CAS: 111771-08-5 | C7H4FIO2 | 266.01 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 111771-08-5
Molecular Formula C7H4FIO2
Molecular Weight (g/mol) 266.01
MDL Number MFCD00042289
InChI Key CYCXAPWOBWWNRK-UHFFFAOYSA-N
Synonym 6-fluoro-2-iodobenzoic acid, benzoic acid, 2-fluoro-6-iodo, maybridge3_007452, pubchem1386, acmc-2099an, 2-fluoro-6-iodobenzoicacid, 2-fluoro-6-iodo benzoic acid, 2-fluoro-6-iodo-benzoic acid, rarechem al bo 0280, attercop-chm at126295
PubChem CID 2733302
IUPAC Name 2-fluoro-6-iodobenzoic acid
SMILES C1=CC(=C(C(=C1)I)C(=O)O)F

Specifications

Melting Point 122°C to 126°C
Quantity 1 g
Beilstein 4982371
Sensitivity Light sensitive
Formula Weight 266.01
Percent Purity 97%
Chemical Name or Material 2-Fluoro-6-iodobenzoic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only