missing translation for 'onlineSavingsMsg'
Learn More

2-Fluoro-5-(trifluoromethyl)pyridine 98.0+%, TCI America™

Catalog No. f08461g
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F08461G 1 g
F08465G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F08461G Supplier TCI America Supplier No. F08461G
Only null left

Chemical Identifiers

CAS 69045-82-5
Molecular Formula C6H3F4N
Molecular Weight (g/mol) 165.091
MDL Number MFCD07437945
InChI Key UUODQIKUTGWMPT-UHFFFAOYSA-N
Synonym 2-fluoro-5-trifluoromethyl pyridine, 2-fluoro-5-trifluoromethyl-pyridine, pyridine, 2-fluoro-5-trifluoromethyl, alpha,alpha,alpha,6-tetrafluoro-3-picoline, pubchem9252, acmc-209o5o, ksc628s4n
PubChem CID 5463754
IUPAC Name 2-fluoro-5-(trifluoromethyl)pyridine
SMILES C1=CC(=NC=C1C(F)(F)F)F

Specifications

Color Yellow
Boiling Point 120°C
Quantity 1 g
UN Number 1993
Formula Weight 165.09
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Fluoro-5-(trifluoromethyl)pyridine